Sodium-1-propanethiolate tech. 85%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

H66135-06 H66135-14
AAH66135-06EA 87.85 USD
AAH66135-06 AAH66135-14
Sodium-1-propanethiolate tech. 85%, Technical Grade
Sodium-1-propanethiolate

Sodium 1-propanethiolate, 95%


Purity: 95% CAS No.: 6898-84-6 Molecular Formula: C3H7NaS Formula Weight: 98.14 Physical Form: Solid Sensitivity: Air and Moisture Sensitive Appearance: White to pale gray Odor: Stench Beilstein Reference: 3556024 UN No.: 3263 MDL No.: MFCD00043318

Formula: C₃H₇NaS
MW: 98.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043318
CAS Number: 6898-84-6
EINECS: 230-005-5
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR