2-Amino-4-methoxypyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H66125-06 H66125-14
AAH66125-06EA 104 USD
AAH66125-06 AAH66125-14
2-Amino-4-methoxypyridine 95%
2-Amino-4-methoxypyridine

2-Amino-4-methoxypyridine, 95%


Purity: 95% CAS No.: 10201-73-7 Molecular Formula: C6H8N2O Formula Weight: 124.14 Physical Form: Solid Sensitivity: Air Sensitive Appearance: Pale yellow Melting Point: 115-118° Beilstein Reference: 471837 UN No.: 2811 MDL No.: MFCD07437849

Formula: C₆H₈N₂O
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD07437849
CAS Number: 10201-73-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR