1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H66108-03
H66108-MD
AAH66108-03EA
385
USD
AAH66108-03
AAH66108-MD
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene 97%
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%
Purity: 97%
CAS No.: 3849-01-2
Molecular Formula: C12H15Cl3
Formula Weight: 265.60
Physical Form: Powder
Appearance: White
Beilstein Reference: 2369523
UN No.: 3261
MDL No.: MFCD00229016
Formula:
C₁₂H₁₅Cl₃ Storage Temperature: Ambient |
MDL Number:
MFCD00229016 CAS Number: 3849-01-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...