tert-Butyl (4-aminocyclohexyl)carbamate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Boc-1,4-diaminocyclohexane
H64932-03
H64932-06
AAH64932-03EA
122
USD
AAH64932-03
AAH64932-06
AAH64932-MD
tert-Butyl (4-aminocyclohexyl)carbamate 95%
tert-Butyl (4-aminocyclohexyl)carbamate
N-Boc-1,4-diaminocyclohexane, 95%
Purity: 95%
CAS No.: 195314-59-1
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
Physical Form: Solid
Appearance: White
Beilstein Reference: 13428745
UN No.: 3259
MDL No.: MFCD01076211
Formula:
C₁₁H₂₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01076211 CAS Number: 195314-59-1 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...