5-Chloropicolinonitrile 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chloro-2-cyanopyridine
H64846-03
H64846-06
AAH64846-03EA
40.4
USD
AAH64846-03
AAH64846-06
AAH64846-14
5-Chloropicolinonitrile 96%
5-Chloropicolinonitrile
5-Chloro-2-cyanopyridine, 96%
Purity: 96%
CAS No.: 89809-64-3
Molecular Formula: C6H3ClN2
Formula Weight: 138.55
Physical Form: Solid
Appearance: Off-white
Melting Point: 106-108°
Beilstein Reference: 113764
UN No.: 3439
MDL No.: MFCD03788835
Formula:
C6H3ClN2 MW: 138.55 g/mol Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788835 CAS Number: 89809-64-3 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...