4-Chloro-2-(trifluoromethyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Chloro-(2-trifluoromethyl)pyridine

H64833-03 H64833-06
AAH64833-03EA 76.9 USD
AAH64833-03 AAH64833-06 AAH64833-MD
4-Chloro-2-(trifluoromethyl)pyridine 98%
4-Chloro-2-(trifluoromethyl)pyridine

4-Chloro-2-(trifluoromethyl)pyridine, 98%


Purity: 98% CAS No.: 131748-14-6 Molecular Formula: C6H3ClF3N Formula Weight: 181.54 Physical Form: Liquid Appearance: Colorless UN No.: 2810 MDL No.: MFCD07368047

Formula: C₆H₃ClF₃N
MW: 181.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368047
CAS Number: 131748-14-6
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR