2,2,2,3'-Tetrafluoroacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2,2-Trifluoro-1-(3-fluorophenyl)ethanone
H64704-03
H64704-06
AAH64704-03EA
30.6
USD
AAH64704-03
AAH64704-06
2,2,2,3'-Tetrafluoroacetophenone 98%
2,2,2,3'-Tetrafluoroacetophenone
2,2,2,3'-Tetrafluoroacetophenone, 98%
Purity: 98%
CAS No.: 708-64-5
Molecular Formula: C8H4F4O
Formula Weight: 192.11
Storage Temperature: 2°C to 8°C
Flash Point: 17°(63°F)
Physical Form: Liquid
Density: 1.372
Refractive Index: 1.4440
Appearance: Colorless to pale yellow
Boiling Point: 79°/50mm
Beilstein Reference: 1872788
UN No.: 1224
MDL No.: MFCD00039236
Formula:
C₈H₄F₄O MW: 192.11 g/mol Boiling Pt: 79 °C/50 mm Density: 1.372 Flash Pt: 17 °C(63 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039236 CAS Number: 708-64-5 EINECS: 211-903-6 UN: 1224 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...