(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-4-Boc-morpholine-3-carboxylic acid
H64654-03
H64654-MD
AAH64654-03EA
299
USD
AAH64654-03
AAH64654-MD
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 97%
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
(R)-4-Boc-morpholine-3-carboxylic acid, 97%
Purity: 97%
CAS No.: 869681-70-9
Molecular Formula: C10H17NO5
Formula Weight: 231.25
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 181°
Beilstein Reference: 10254221
UN No.: 2811
MDL No.: MFCD04114909
Formula:
C10H17NO5 MW: 231.25 g/mol Melting Pt: 181 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114909 CAS Number: 869681-70-9 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...