2,4-Dichloro-6-nitrophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64524-14
H64524-22
AAH64524-14EA
86.7
USD
AAH64524-14
AAH64524-22
2,4-Dichloro-6-nitrophenol 98+%
2,4-Dichloro-6-nitrophenol
2,4-Dichloro-6-nitrophenol, 98+%
Purity: 98+%
CAS No.: 609-89-2
Molecular Formula: C6H3Cl2NO3
Formula Weight: 207.99
Physical Form: Powder
Appearance: Yellow
Melting Point: 118-120°
Beilstein Reference: 2050081
UN No.: 2811
MDL No.: MFCD00007101
Formula:
C₆H₃Cl₂NO₃ MW: 208 g/mol Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007101 CAS Number: 609-89-2 EINECS: 210-202-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...