2-Bromo-3-(trifluoromethyl)aniline 98%

Supplier: Thermo Scientific Chemicals
Warning

H64309-03 H64309-06
AAH64309-03EA 27.6 USD
AAH64309-03 AAH64309-06
2-Bromo-3-(trifluoromethyl)aniline 98%
2-Bromo-3-(trifluoromethyl)aniline

2-Bromo-3-(trifluoromethyl)aniline, 98%


Purity: 98% CAS No.: 58458-10-9 Molecular Formula: C7H5BrF3N Formula Weight: 240.02 Physical Form: Solid Appearance: Off-white Beilstein Reference: 2835356 UN No.: 2811 MDL No.: MFCD01656033

Formula: C₇H₅BrF₃N
MW: 240.02
Storage Temperature: Ambient
MDL Number: MFCD01656033
CAS Number: 58458-10-9
EINECS: 261-261-6
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR