6-fluoronicotinonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Cyano-2-fluoropyridine, 6-Fluoronicotinonitrile
H64278-03
H64278-06
AAH64278-03EA
63.4
USD
AAH64278-03
AAH64278-06
AAH64278-MD
6-fluoronicotinonitrile 95%
6-fluoronicotinonitrile
5-Cyano-2-fluoropyridine, 95%
Purity: 95%
CAS No.: 3939-12-6
Molecular Formula: C6H3FN2
Formula Weight: 122.10
Physical Form: Solid
Appearance: Off-white
Melting Point: 51-52°
Beilstein Reference: 471855
UN No.: 3439
MDL No.: MFCD09261087
Formula:
C₆H₃FN₂ MW: 122.102 g/mol Melting Pt: 51…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD09261087 CAS Number: 3939-12-6 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...