N-Methyl-3-(trifluoromethyl)benzylamine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H63888-03
H63888-06
AAH63888-03EA
94
USD
AAH63888-03
AAH63888-06
N-Methyl-3-(trifluoromethyl)benzylamine 95%
N-Methyl-3-(trifluoromethyl)benzylamine
N-Methyl-3-(trifluoromethyl)benzylamine, 95%
Purity: 95%
CAS No.: 90390-07-1
Molecular Formula: C9H10F3N
Formula Weight: 189.18
Physical Form: Oil
Appearance: Colorless
UN No.: 2735
MDL No.: MFCD04115410
Formula:
C₉H₁₀F₃N Storage Temperature: Ambient |
MDL Number:
MFCD04115410 CAS Number: 90390-07-1 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...