6-Chloro-4-methylpyridine-2-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

H63584-03 H63584-06
AAH63584-03EA 108 USD
AAH63584-03 AAH63584-06 AAH63584-MD
6-Chloro-4-methylpyridine-2-carboxylic acid 95%
6-Chloro-4-methylpyridine-2-carboxylic acid

6-Chloro-4-methylpyridine-2-carboxylic acid, 95%


Purity: 95% CAS No.: 324028-95-7 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Physical Form: Solid Appearance: Off-white UN No.: 2811 MDL No.: MFCD09834827

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834827
CAS Number: 324028-95-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR