4-[2-(Boc-amino)ethyl]aniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl 4-aminophenethylcarbamate, 4-(2-Aminoethyl)aniline, 4-BOC protected
H63324-03
H63324-06
AAH63324-03EA
87.4
USD
AAH63324-03
AAH63324-06
4-[2-(Boc-amino)ethyl]aniline 97%
4-[2-(Boc-amino)ethyl]aniline
4-[2-(Boc-amino)ethyl]aniline, 97%
Purity: 97%
CAS No.: 94838-59-2
Molecular Formula: C13H20N2O2
Formula Weight: 236.31
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Brown
UN No.: 2811
MDL No.: MFCD05663961
Formula:
C₁₃H₂₀N₂O₂ MW: 236.315 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663961 CAS Number: 94838-59-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...