4-Fluoro-3-nitrotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63317-14
H63317-22
AAH63317-14EA
81.3
USD
AAH63317-14
AAH63317-22
4-Fluoro-3-nitrotoluene 98%
4-Fluoro-3-nitrotoluene
4-Fluoro-3-nitrotoluene, 98%
Purity: 98%
CAS No.: 446-11-7
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Physical Form: Liquid
Appearance: Red-brown
Boiling Point: 241° dec
Beilstein Reference: 2253853
UN No.: 2810
MDL No.: MFCD00007060
Formula:
C₇H₆FNO₂ Boiling Pt: 241° (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007060 CAS Number: 446-11-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...