tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63191-03
H63191-06
AAH63191-03EA
94.79
USD
AAH63191-03
AAH63191-06
AAH63191-MD
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 97%
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate
1-Boc-4-(4-aminophenyl)piperazine, 97%
Purity: 97%
CAS No.: 170911-92-9
Molecular Formula: C15H23N3O2
Formula Weight: 277.36
Physical Form: Solid
MDL No.: MFCD04115065
Formula:
C15H23N3O2 Storage Temperature: Ambient |
MDL Number:
MFCD04115065 CAS Number: 170911-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...