2-Fluoro-3-nitrotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-1-methyl-3-nitrobenzene

H63173-14 H63173-22
AAH63173-14EA 92.6 USD
AAH63173-14 AAH63173-22
2-Fluoro-3-nitrotoluene 98%
2-Fluoro-3-nitrotoluene

2-Fluoro-3-nitrotoluene, 98%


Purity: 98% CAS No.: 437-86-5 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Flash Point: 98°(208°F) Physical Form: Liquid Density: 1.276 Refractive Index: 1.5290 Appearance: Colorless to pale yellow Melting Point: 19° dec. Boiling Point: 111°/12mm Beilstein Reference: 2442082 UN No.: 2810 MDL No.: MFCD03412242

Formula: C₇H₆FNO₂
Boiling Pt: 111 °C/12 mm
Melting Pt: 19 °C (dec.)
Density: 1.276
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD03412242
CAS Number: 437-86-5
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR