6-(Trifluoromethyl)picolinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-(Trifluoromethyl)-2-pyridinecarboxylic acid, 6-(Trifluoromethyl)picolinic acid, 2-(Trifluoromethyl)-6-pyridinecarboxylic acid, 6-(Trifluoromethyl)pyridine-2-carboxylic acid
H63138-03
H63138-MD
AAH63138-03EA
113
USD
AAH63138-03
AAH63138-MD
6-(Trifluoromethyl)picolinic acid 98%
6-(Trifluoromethyl)picolinic acid
6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%
Purity: 98%
CAS No.: 131747-42-7
Molecular Formula: C7H4F3NO2
Formula Weight: 191.11
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 154-158°
UN No.: 2811
MDL No.: MFCD02180819
Formula:
C₇H₄F₃NO₂ MW: 191.11 g/mol Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD02180819 CAS Number: 131747-42-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...