3'-Bromo-5'-fluoroacetophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3-Bromo-5-fluorophenyl)ethanone

H62977-03 H62977-06
AAH62977-03EA 138 USD
AAH62977-03 AAH62977-06 AAH62977-MD
3'-Bromo-5'-fluoroacetophenone 96%
3'-Bromo-5'-fluoroacetophenone

3'-Bromo-5'-fluoroacetophenone, 96%


Purity: 96% CAS No.: 105515-20-6 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847746
CAS Number: 105515-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR