Boc-D-4-Cyanophenylalanine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-D-Phe(4-CN)-OH, N-Boc-4-cyano-D-phenylalanine, Boc-4-cyano-D-Phe-OH

H62947-03 H62947-06 H62947-MD
AAH62947-03EA 126 USD
AAH62947-03 AAH62947-06 AAH62947-MD
Boc-D-4-Cyanophenylalanine 98%
Boc-D-4-Cyanophenylalanine

N-Boc-4-cyano-D-phenylalanine, 98%


Purity: 98% CAS No.: 146727-62-0 Molecular Formula: C15H18N2O4 Formula Weight: 290.31 Beilstein Reference: 9339089 UN No.: 3439 MDL No.: MFCD00672527

Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672527
CAS Number: 146727-62-0
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR