2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Carbamoyl-3-fluorophenylboronic acid pinacol ester
H62904-03
H62904-MD
AAH62904-03EA
478
USD
AAH62904-03
AAH62904-MD
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester, 96%
Purity: 96%
Molecular Formula: C13H17BFNO3
Formula Weight: 265.09
Physical Form: Solid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...