3'-Bromo-2'-fluoroacetophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

H62708-03 H62708-MD
AAH62708-03EA 173 USD
AAH62708-03 AAH62708-06 AAH62708-MD
3'-Bromo-2'-fluoroacetophenone 96%
3'-Bromo-2'-fluoroacetophenone

3-Bromo-2-fluoroacetophenone, 96%


Purity: 96% CAS No.: 161957-61-5 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD09264507
CAS Number: 161957-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR