Methyl-3-bromo-2-fluorophenylacetate 96%
Supplier: Thermo Scientific Chemicals
H62655-03
H62655-06
AAH62655-03EA
227
USD
AAH62655-03
AAH62655-06
AAH62655-MD
Methyl-3-bromo-2-fluorophenylacetate 96%
Methyl-3-bromo-2-fluorophenylacetate
Methyl 3-Bromo-2-fluorophenylacetate, 96%
Purity: 96%
Molecular Formula: C9H8BrFO2
Formula Weight: 247.06
Physical Form: Solid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...