Methyl (3-bromo-5-fluorophenyl)acetate 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 2-(3-bromo-5-fluorophenyl)acetate
H62561-03
H62561-06
AAH62561-03EA
196
USD
AAH62561-03
AAH62561-06
AAH62561-MD
Methyl (3-bromo-5-fluorophenyl)acetate 96%
Methyl (3-bromo-5-fluorophenyl)acetate
Methyl 3-bromo-5-fluorophenylacetate, 96%
Purity: 96%
CAS No.: 937732-38-2
Molecular Formula: C9H8BrFO2
Formula Weight: 247.06
Physical Form: Solid
Formula:
C₉H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD20489056 CAS Number: 937732-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...