2-Amino-4-fluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzenamine, 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 2-Amino-4-fluorophenylboronic acid pinacol ester, 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

H62511-03 H62511-MD
AAH62511-03EA 616 USD
AAH62511-03 AAH62511-MD
2-Amino-4-fluorobenzeneboronic acid pinacol ester 96%
2-Amino-4-fluorobenzeneboronic acid pinacol ester

2-Amino-4-fluorobenzeneboronic acid pinacol ester, 96%


Purity: 96% Molecular Formula: C12H17BFNO2 Formula Weight: 237.08 Physical Form: Solid

Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD12025504
CAS Number: 1309982-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR