Methyl-4-bromo-2-fluorophenylacetate 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-(4-bromo-2-fluorophenyl)acetate

H62482-03 H62482-MD
AAH62482-03EA 190 USD
AAH62482-03 AAH62482-06 AAH62482-MD
Methyl-4-bromo-2-fluorophenylacetate 96%
Methyl-4-bromo-2-fluorophenylacetate

Methyl 4-bromo-2-fluorophenylacetate, 96%


Purity: 96% CAS No.: 193290-19-6 Molecular Formula: C9H8BrFO2 Formula Weight: 247.06 Physical Form: Solid

Formula: C₉H₈BrFO₂
MW: 247.063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483545
CAS Number: 193290-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR