Di-tert-butyl-L-(+)-tartrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R)-Di-tert-butyl 2,3-dihydroxysuccinate, L-(+)-Tartaric Acid Di-tert-Butyl Ester
H55860-03
H55860-MD
AAH55860-03EA
331
USD
AAH55860-03
AAH55860-MD
AAH55860-ME
Di-tert-butyl-L-(+)-tartrate 99%
Di-tert-butyl-L-(+)-tartrate
(+)-Di-tert-butyl L-tartrate, 99%
Purity: 99%
CAS No.: 117384-45-9
Molecular Formula: C12H22O6
Formula Weight: 262.30
Flash Point: 109°(228°F)
Physical Form: Solid
Density: 1.143
Appearance: White
Melting Point: 90-92°
Beilstein Reference: 5026723
Optical Rotation: +11° (c=1 in acetone)
MDL No.: MFCD00192000
Formula:
C₁₂H₂₂O₆ Melting Pt: 90…92 °C Density: 1.143 Flash Pt: 109 °C(228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00192000 CAS Number: 117384-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...