1-(1-Naphthylmethyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Naphthalen-1-ylmethyl)piperazine

H55398-03 H55398-06 H55398-14
AAH55398-03EA 64.94 USD
AAH55398-03 AAH55398-06 AAH55398-14
1-(1-Naphthylmethyl)piperazine 97%
1-(1-Naphthylmethyl)piperazine

1-(1-Naphthylmethyl)piperazine, 97%


Purity: 97% CAS No.: 40675-81-8 Molecular Formula: C15H18N2 Formula Weight: 226.32 Physical Form: Powder Appearance: White to brown Melting Point: 63-67° UN No.: 3077 MDL No.: MFCD01314185

Formula: C₁₅H₁₈N₂
MW: 226.323 g/mol
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01314185
CAS Number: 40675-81-8
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR