4-Boc-1-(5-bromo-2-pyridyl)piperazine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Bromo-2-(4-tert-butoxycarbonylpiperazin-1-yl)pyridine, tert-Butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
H55311-03
H55311-06
H55311-14
AAH55311-03EA
41.1
USD
AAH55311-03
AAH55311-06
AAH55311-14
4-Boc-1-(5-bromo-2-pyridyl)piperazine 97%
4-Boc-1-(5-bromo-2-pyridyl)piperazine
4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%
Purity: 97%
CAS No.: 153747-97-8
Molecular Formula: C14H20BrN3O2
Formula Weight: 342.23
Physical Form: Powder
Appearance: White to yellow
Melting Point: 83-87°
UN No.: 2811
MDL No.: MFCD07369772
Formula:
C₁₄H₂₀BrN₃O₂ MW: 342.237 g/mol Melting Pt: 83…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD07369772 CAS Number: 153747-97-8 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...