Palladium 5% on activated carbon, powder Type 487

Supplier: Thermo Scientific Chemicals
Warning

H37587-06 H37587-14
AAH37587-06EA 100 USD
AAH37587-06 AAH37587-14
Palladium 5% on activated carbon, powder Type 487
Palladium

Palladium, 5% on activated carbon powder, Type 487


CAS No.: 7440-05-3 Physical Form: Powder Appearance: Dark grey Odor: Odorless UN No.: 1362 MDL No.: MFCD03457879

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 1362
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR