Palladium ≥99.95% (metals basis), powder , APS 0.35-0.8 micron
Supplier: Thermo Scientific Chemicals
|
Danger
|
047249-03
047249-06
047249-77
AA47249-03EA
331
USD
AA47249-03
AA47249-06
AA47249-77
Palladium ≥99.95% (metals basis), powder , APS 0.35-0.8 micron
Palladium
Palladium powder, APS 0.35-0.8 micron, 99.95% (metals basis)
Purity: 99.95%
CAS No.: 7440-05-3
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011167
Formula:
Pd MW: 106.420 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 UN: 3089 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...