2'-Bromo-6'-fluoroacetophenone 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2-Bromo-6-fluorophenyl)ethanone
H62361-03
H62361-06
H62361-MD
AAH62361-03EA
190
USD
AAH62361-03
AAH62361-06
AAH62361-MD
2'-Bromo-6'-fluoroacetophenone 96%
2'-Bromo-6'-fluoroacetophenone
2'-Bromo-6'-fluoroacetophenone, 96%
Purity: 96%
CAS No.: 928715-37-1
Molecular Formula: C8H6BrFO
Formula Weight: 217.04
Physical Form: Solid
Formula:
C₈H₆BrFO MW: 217.04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13193629 CAS Number: 928715-37-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...