2-Bromo-4-fluorobenzylamine hydrochloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Bromo-4-fluorophenyl)methanamine hydrochloride
H62200-03
H62200-06
AAH62200-03EA
185
USD
AAH62200-03
AAH62200-06
AAH62200-MD
2-Bromo-4-fluorobenzylamine hydrochloride 96%
2-Bromo-4-fluorobenzylamine hydrochloride
2-Bromo-4-fluorobenzylamine hydrochloride, 96%
Purity: 96%
CAS No.: 289038-14-8
Molecular Formula: C7H7BrFN.HCl
Formula Weight: 240.50
Physical Form: Solid
MDL No.: MFCD00672922
Formula:
C₇H₈BrClFN Storage Temperature: Ambient |
MDL Number:
MFCD00672922 CAS Number: 289038-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...