4'-Bromo-2'-fluoroacetophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Bromo-2-fluorophenyl)ethanone

H62127-03 H62127-06
AAH62127-03EA 185 USD
AAH62127-03 AAH62127-06 AAH62127-MD
4'-Bromo-2'-fluoroacetophenone 96%
4'-Bromo-2'-fluoroacetophenone

4'-Bromo-2'-fluoroacetophenone, 96%


Purity: 96% CAS No.: 625446-22-2 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411551
CAS Number: 625446-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR