2-Bromo-6-fluorobenzylamine hydrochloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2-Bromo-6-fluorophenyl)methanamine hydrochloride (1:1), Benzenemethanamine, 2-bromo-6-fluoro-, hydrochloride (1:1)
H62086-03
H62086-06
AAH62086-03EA
176
USD
AAH62086-03
AAH62086-06
AAH62086-MD
2-Bromo-6-fluorobenzylamine hydrochloride 96%
2-Bromo-6-fluorobenzylamine hydrochloride
2-Bromo-6-fluorobenzylamine hydrochloride, 96%
Purity: 96%
CAS No.: 1214330-90-1
Molecular Formula: C7H7BrFN.HCl
Formula Weight: 240.50
Physical Form: Solid
Formula:
C₇H₇BrFN·HCl Storage Temperature: Ambient |
MDL Number:
MFCD12546112 CAS Number: 1214330-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...