4'-Bromo-3'-fluoroacetophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

H62040-06 H62040-MD
AAH62040-03EA 174.27 USD
AAH62040-03 AAH62040-06 AAH62040-MD
4'-Bromo-3'-fluoroacetophenone 96%
4'-Bromo-3'-fluoroacetophenone

4-Bromo-3-fluoroacetophenone, 96%


Purity: 96% CAS No.: 304445-49-6 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD00466241
CAS Number: 304445-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR