Boc-Phe(4-CN)-OH 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Boc-4-cyano-Phe-OH, N-Boc-4-cyano-L-phenylalanine, Boc-Phe(4-CN)-OH, N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine
H62023-03
H62023-06
AAH62023-03EA
115
USD
AAH62023-03
AAH62023-06
Boc-Phe(4-CN)-OH 95%
Boc-Phe(4-CN)-OH
N-Boc-4-cyano-L-phenylalanine, 95%
Purity: 95%
CAS No.: 131724-45-3
Molecular Formula: C15H18N2O4
Formula Weight: 290.31
Physical Form: Powder
Appearance: White
Melting Point: 156-160°
Beilstein Reference: 7715793
UN No.: 3439
MDL No.: MFCD00672526
Formula:
C₁₅H₁₈N₂O₄ MW: 290.319 g/mol Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672526 CAS Number: 131724-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...