2-Fluoro-4-iodobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
2-Fluoro-4-iodobenzonitrile, 98%
Purity: 98%
CAS No.: 137553-42-5
Molecular Formula: C7H3FIN
Formula Weight: 247.01
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Red-brown
UN No.: 3439
MDL No.: MFCD03094172
Formula:
C₇H₃FIN Storage Temperature: Ambient |
MDL Number:
MFCD03094172 CAS Number: 137553-42-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...