2-Bromo-3-fluoroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H61687-03 H61687-06
AAH61687-03EA 89.3 USD
AAH61687-03 AAH61687-06 AAH61687-MD
2-Bromo-3-fluoroaniline 97%
2-Bromo-3-fluoroaniline

2-Bromo-3-fluoroaniline, 97%


Purity: 97% CAS No.: 111721-75-6 Molecular Formula: C6H5BrFN Formula Weight: 190.01 Flash Point: 94°(201°F) Physical Form: Crystalline Density: 1.670 Refractive Index: 1.5830 Appearance: Off-white UN No.: 2811 MDL No.: MFCD07369915

Formula: C₆H₅BrFN
Density: 1.670
Flash Pt: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD07369915
CAS Number: 111721-75-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR