α,3-Dibromo-4-fluorotoluene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Bromo-4-fluorobenzyl bromide, 2-Bromo-4-(bromomethyl)-1-fluorobenzene
H61399-06
H61399-14
H61399-22
AAH61399-06EA
33.2
USD
AAH61399-06
AAH61399-14
AAH61399-22
α,3-Dibromo-4-fluorotoluene 97%
α,3-Dibromo-4-fluorotoluene
3-Bromo-4-fluorobenzyl bromide, 97%
Purity: 97%
CAS No.: 78239-71-1
Molecular Formula: C7H5Br2F
Formula Weight: 267.92
Physical Form: Solid
Appearance: White
Melting Point: 41°
UN No.: 3261
MDL No.: MFCD09037367
Formula:
C₇H₅Br₂F Melting Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037367 CAS Number: 78239-71-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...