2-Fluoro-m-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-3-methylaniline, 3-Amino-2-fluorotoluene
H61143-14
H61143-22
AAH61143-14EA
162
USD
AAH61143-14
AAH61143-22
2-Fluoro-m-toluidine 98%
2-Fluoro-m-toluidine
2-Fluoro-3-methylaniline, 98%
Purity: 98%
CAS No.: 1978-33-2
Molecular Formula: C7H8FN
Formula Weight: 125.14
Refractive Index: 1.5360
Boiling Point: 87°/12mm
UN No.: 2810
MDL No.: MFCD06410915
Formula:
C₇H₈FN MW: 125.14 g/mol Boiling Pt: 87 °C/12 mm Storage Temperature: Ambient |
MDL Number:
MFCD06410915 CAS Number: 1978-33-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...