α-Bromo-4-chlorophenylacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H60141-03
H60141-06
AAH60141-03EA
41.7
USD
AAH60141-03
AAH60141-06
α-Bromo-4-chlorophenylacetic acid 97%
α-Bromo-4-chlorophenylacetic acid
^a-Bromo-4-chlorophenylacetic acid, 97%
Purity: 97%
CAS No.: 3381-73-5
Molecular Formula: C8H6BrClO2
Formula Weight: 249.49
Melting Point: 95-99°
UN No.: 3261
MDL No.: MFCD08276760
Formula:
C₈H₆BrClO₂ MW: 249.49 g/mol Melting Pt: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276760 CAS Number: 3381-73-5 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...