Methyl-N-acetyl-L-tryptophanate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Acetyl-L-tryptophan methyl ester, Methyl N-acetyl-L-tryptophanate, Ac-Trp-OMe

H66937-06 H66937-14
AAH66937-06EA 74.4 USD
AAH66937-06 AAH66937-14
Methyl-N-acetyl-L-tryptophanate 95%
Methyl-N-acetyl-L-tryptophanate

N-Acetyl-L-tryptophan methyl ester, 95%


Purity: 95% CAS No.: 2824-57-9 Molecular Formula: C14H16N2O3 Formula Weight: 260.29 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white to white MDL No.: MFCD02728656

Formula: C₁₄H₁₆N₂O₃
MW: 260.293 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02728656
CAS Number: 2824-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR