N-Boc-N-methyl-1,3-diaminopropane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(N-Boc-N-methylamino)propylamine, N-Methylpropane-1,3-diamine, N-BOC protected
H66932-03
H66932-MD
AAH66932-03EA
141
USD
AAH66932-03
AAH66932-MD
N-Boc-N-methyl-1,3-diaminopropane 95%
N-Boc-N-methyl-1,3-diaminopropane
N-Boc-N-methyl-1,3-diaminopropane, 95%
Purity: 95%
CAS No.: 150349-36-3
Molecular Formula: C9H20N2O2
Formula Weight: 188.27
Physical Form: Liquid
Appearance: Pale yellow
MDL No.: MFCD01631189
Formula:
C₉H₂₀N₂O₂ MW: 188.271 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631189 CAS Number: 150349-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...