Fmoc-D-Lys(Alloc)-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nepsilon-Allyloxycarbonyl-Nα-Fmoc-D-lysine, Fmoc-D-Lys(Aloc)-OH
H66861-03
H66861-06
AAH66861-03EA
42
USD
AAH66861-03
AAH66861-06
Fmoc-D-Lys(Alloc)-OH 95%
Fmoc-D-Lys(Alloc)-OH
N^e-Allyloxycarbonyl-N^a-Fmoc-D-lysine, 95%
Purity: 95%
CAS No.: 214750-75-1
Molecular Formula: C25H28N2O6
Formula Weight: 452.51
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00798639
Formula:
C₂₅H₂₈N₂O₆ MW: 452.507 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798639 CAS Number: 214750-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...