N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Z-Tyr(tBu)-OH
H66762-06
H66762-22
AAH66762-06EA
42.9
USD
AAH66762-06
AAH66762-14
AAH66762-22
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine, 98%
Purity: 98%
CAS No.: 5545-54-0
Molecular Formula: C21H25NO5
Formula Weight: 371.43
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 2781067
MDL No.: MFCD00065701
Formula:
C₂₁H₂₅NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00065701 CAS Number: 5545-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...