Boc-D-asp-Otbu 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-4-(tert-Butoxy)-3-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid, N-Boc-D-aspartic acid 1-tert-butyl ester

H66716-03 H66716-06
AAH66716-03EA 63.4 USD
AAH66716-03 AAH66716-06 AAH66716-MD
Boc-D-asp-Otbu 98%
Boc-D-asp-Otbu

N-Boc-D-aspartic acid 1-tert-butyl ester, 98%


Purity: 98% CAS No.: 77004-75-2 Molecular Formula: C13H23NO6 Formula Weight: 289.33 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 3553120 MDL No.: MFCD00236827

Formula: C₁₃H₂₃NO₆
MW: 289.328 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236827
CAS Number: 77004-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR