N-Acetyl-S-trityl-L-cysteine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ac-Cys(Trt)-OH
H66689-03
H66689-06
AAH66689-03EA
98.6
USD
AAH66689-03
AAH66689-06
N-Acetyl-S-trityl-L-cysteine 95%
N-Acetyl-S-trityl-L-cysteine
N-Acetyl-S-trityl-L-cysteine, 95%
Purity: 95%
CAS No.: 27486-87-9
Molecular Formula: C24H23NO3S
Formula Weight: 405.51
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 6877409
MDL No.: MFCD00236747
Formula:
C₂₄H₂₃NO₃S Storage Temperature: Refrigerator |
MDL Number:
MFCD00236747 CAS Number: 27486-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...