4-Chloropyrimidin-5-amine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
5-Amino-4-chloropyrimidine, 96%
Purity: 96%
CAS No.: 54660-78-5
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Appearance: White to pale brown
Melting Point: 110-116°
MDL No.: MFCD06412561
Formula:
C4H4ClN3 Melting Pt: 110…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD06412561 CAS Number: 54660-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...