Fmoc-Arg-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nα-Fmoc-L-arginine
H66665-06
H66665-14
AAH66665-06EA
53.58
USD
AAH66665-06
AAH66665-14
Fmoc-Arg-OH 95%
Fmoc-Arg-OH
N^a-Fmoc-L-arginine, 95%
Purity: 95%
CAS No.: 91000-69-0
Molecular Formula: C21H24N4O4
Formula Weight: 396.45
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Off-white
Beilstein Reference: 4828015
MDL No.: MFCD00051770
Formula:
C₂₁H₂₄N₄O₄ MW: 396.447 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00051770 CAS Number: 91000-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...